Learn More
Methyl 3,3-dimethylacrylate, 98%
CAS: 924-50-5 | C6H10O2 | 114.144 g/mol
$142.47 - $492.52
Chemical Identifiers
| CAS | 924-50-5 |
|---|---|
| Molecular Formula | C6H10O2 |
| Molecular Weight (g/mol) | 114.144 |
| MDL Number | MFCD00043940 |
| InChI Key | FZIBCCGGICGWBP-UHFFFAOYSA-N |
| Synonym | methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate |
| PubChem CID | 13546 |
| IUPAC Name | methyl 3-methylbut-2-enoate |
| SMILES | CC(=CC(=O)OC)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1973109
|
Thermo Scientific Chemicals
L1973109 |
10 g |
Each for $142.47
|
|
|||||
|
AAL1973118
|
Thermo Scientific Chemicals
L1973118 |
50 g |
Each for $492.52
|
|
|||||
Description
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in chloroform. Insoluble in water,
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.
Chemical Identifiers
| 924-50-5 | |
| 114.144 | |
| FZIBCCGGICGWBP-UHFFFAOYSA-N | |
| 13546 | |
| CC(=CC(=O)OC)C |
| C6H10O2 | |
| MFCD00043940 | |
| methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate | |
| methyl 3-methylbut-2-enoate |
Specifications
| 924-50-5 | |
| 0.936 | |
| 33°C (91°F) | |
| 1.4375 | |
| MFCD00043940 | |
| UN3272 | |
| Light sensitive | |
| Soluble in chloroform. Insoluble in water, | |
| CC(=CC(=O)OC)C | |
| 114.144 | |
| 114.14 | |
| Methyl 3,3-dimethylacrylate |
| -41°C | |
| 134°C to 139°C | |
| C6H10O2 | |
| (CH3)2C=CHCO2CH3 | |
| 10 g | |
| 1741592 | |
| methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate | |
| FZIBCCGGICGWBP-UHFFFAOYSA-N | |
| methyl 3-methylbut-2-enoate | |
| 13546 | |
| 98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 213-107-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only