missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-morpholin-4-ylethoxy)benzoate, 97%, Thermo Scientific™

Catalog No. CC42022DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC42022DA 1g
CC42022DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC42022DA Supplier Thermo Scientific Chemicals Supplier No. CC42022DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 249937-00-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD08060518
InChI Key VOYSHDFODOJUMP-UHFFFAOYSA-N
Synonym methyl 3-2-morpholin-4-ylethoxy benzoate, methyl 3-2-morpholin-4-yl ethoxy benzoate, methyl 3-2-morpholinoethoxy benzoate, methyl 3-2-morpholin4-ylethoxy benzoate, 3-2-morpholin-4-yl-ethoxy-benzoic acid methyl ester, benzoic acid,3-2-4-morpholinyl ethoxy-, methyl ester
PubChem CID 7164628
IUPAC Name methyl 3-(2-morpholin-4-ylethoxy)benzoate
SMILES COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material Methyl 3-(2-morpholin-4-ylethoxy)benzoate
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.