missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-[(2-Methoxy-2-oxoethyl)thio]propionate 98.0+%, TCI America™

Catalog No. p-7133504
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29221G 1 g
M29225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29221G Supplier TCI America Supplier No. M29221G

Chemical Identifiers

CAS 7400-45-5
Molecular Formula C7H12O4S
Molecular Weight (g/mol) 192.229
MDL Number MFCD00043633
InChI Key XNDKLLFGXIEGKL-UHFFFAOYSA-N
Synonym dimethyl 3-thiaadipate, methyl 3-2-methoxy-2-oxoethyl thio propionate, methyl 3-2-methoxy-2-oxoethyl sulfanyl propanoate, methyl 3-methoxycarbonylmethylthio propionate, methyl 3-2-methoxy-2-oxoethyl thio propanoate, propanoic acid, 3-2-methoxy-2-oxoethyl thio-, methyl ester, propanoic acid, 3-carbomethoxymethylthio-, methyl ester, methyl 3-methoxycarbonyl methylthio propanoate, methyl 3-methoxycarbonyl methylthio propionate, 3-2-methoxy-2-oxoethyl thio propionic acid methyl ester
PubChem CID 81891
IUPAC Name methyl 3-(2-methoxy-2-oxoethyl)sulfanylpropanoate
SMILES COC(=O)CCSCC(=O)OC

Specifications

Color Yellow
Boiling Point 117°C
Quantity 1 g
UN Number 3272
Formula Weight 192.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-[(2-Methoxy-2-oxoethyl)thio]propionate