missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-Bromophenyl)propionate 98.0+%, TCI America™

Catalog No. M25871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25871G 1 g
M25875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25871G Supplier TCI America Supplier No. M25871G

Chemical Identifiers

CAS 66191-86-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953149
InChI Key NIULOVGPRAJIJM-UHFFFAOYSA-N
Synonym methyl 3-2-bromophenyl propanoate, methyl 3-2-bromophenyl propionate, benzenepropanoic acid, 2-bromo-, methyl ester, 3-2-bromo-phenyl-propionic acid methyl ester, 3-2-bromophenyl propanoic acid,methyl ester, ksc495a4p, 2-bromohydrocinnamic acid methyl ester, 3-2-bromophenyl propanoic acid methyl ester, 3-2-bromophenyl propionic acid methyl ester
PubChem CID 15712815
IUPAC Name methyl 3-(2-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 87°C
Quantity 1 g
Formula Weight 243.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-(2-Bromophenyl)propionate
TSCA No
Recommended Storage Refrigerator