missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-bromophenyl)propionate, 98%

Catalog No. p-7053718
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6134206 5 g
AAH6134214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6134206 Supplier Thermo Scientific Chemicals Supplier No. H6134206
Only null left

CAS: 66191-86-4 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66191-86-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953149
InChI Key NIULOVGPRAJIJM-UHFFFAOYSA-N
Synonym methyl 3-2-bromophenyl propanoate, methyl 3-2-bromophenyl propionate, benzenepropanoic acid, 2-bromo-, methyl ester, 3-2-bromo-phenyl-propionic acid methyl ester, 3-2-bromophenyl propanoic acid,methyl ester, ksc495a4p, 2-bromohydrocinnamic acid methyl ester, 3-2-bromophenyl propanoic acid methyl ester, 3-2-bromophenyl propionic acid methyl ester
PubChem CID 15712815
IUPAC Name methyl 3-(2-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1Br

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 243.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-(2-bromophenyl)propionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only