missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2S,3S)-2-Isocyanato-3-methylvalerate 98.0+%, TCI America™

Catalog No. I04715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04715G Supplier TCI America Supplier No. I04715G
Only null left

Chemical Identifiers

CAS 120219-17-2
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD00191534
InChI Key OLHBZFKROSWSCI-UHFFFAOYNA-N
Synonym (2S,3S)-2-Isocyanato-3-methylvaleric Acid Methyl Ester
PubChem CID 7019165
IUPAC Name methyl 2-isocyanato-3-methylpentanoate
SMILES CCC(C)C(N=C=O)C(=O)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (2S,3S)-2-Isocyanato-3-methylvalerate