CAS RN 120219-17-2
CAS RN 120219-17-2
IUPAC Name:
methyl 2-isocyanato-3-methylpentanoate
Synonyms:
methyl 2-isocyanato-3-methylpentanoate
Molecular Weight (g/mol):
171.2
Molecular Formula:
C8H13NO3
InChi Key:
OLHBZFKROSWSCI-UHFFFAOYNA-N
SMILES:
CCC(C)C(N=C=O)C(=O)OC
1
–
1
of
1
results
1
Methyl (2S,3S)-2-Isocyanato-3-methylvalerate 98.0+%, TCI America™
CAS: 120219-17-2 Molecular Formula: C8H13NO3 Molecular Weight (g/mol): 171.20 MDL Number: MFCD00191534 InChI Key: OLHBZFKROSWSCI-UHFFFAOYNA-N Synonym: (2S,3S)-2-Isocyanato-3-methylvaleric Acid Methyl Ester PubChem CID: 7019165 IUPAC Name: methyl 2-isocyanato-3-methylpentanoate SMILES: CCC(C)C(N=C=O)C(=O)OC