missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate, 98%

Catalog No. AAH6164103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6164103 1 g
AAH6164106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6164103 Supplier Thermo Scientific Chemicals Supplier No. H6164103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32981-85-4 | C17H17NO4 | 299.326 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32981-85-4
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.326
MDL Number MFCD00673331
InChI Key UYJLJICUXJPKTB-LSDHHAIUSA-N
Synonym 2r,3s-methyl 3-benzamido-2-hydroxy-3-phenylpropanoate, methyl 2r,3s-3-benzoylamino-2-hydroxy-3-phenylpropanoate, bz-rs-iser 3-ph-ome, benzenepropanoic acid, beta-benzoylamino-alpha-hydroxy-, methyl ester, alphar,betas, methyl 2r,3s-3-benzamido-2-hydroxy-3-phenylpropanoate, 2r,3s-n-tert-butoxycarbony-3-phenylisoserine methyl ester, methyl 2r,3s-2-hydroxy-3-phenyl-3-phenylformamido propanoate, pubchem9699, n-benzoyl-2r,3s-3-phenylisoserine methyl ester, methyl 2r,3s-n-benzoylphenylisoserinate
PubChem CID 182104
IUPAC Name methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
SMILES COC(=O)C(C(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2)O

Specifications

Melting Point 178°C to 183°C
Color White
Quantity 1 g
Formula Weight 299.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only