missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(Trifluoromethyl)-3,3,3-trifluoropropionate 98.0+%, TCI America™
$149.13 - $510.75
Chemical Identifiers
| CAS | 360-54-3 |
|---|---|
| Molecular Formula | C5H4F6O2 |
| Molecular Weight (g/mol) | 210.075 |
| MDL Number | MFCD00054663 |
| InChI Key | JGGBVFLRNNDHCM-UHFFFAOYSA-N |
| Synonym | 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester |
| PubChem CID | 9673 |
| IUPAC Name | methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate |
| SMILES | COC(=O)C(C(F)(F)F)C(F)(F)F |
Chemical Identifiers
| 360-54-3 | |
| 210.075 | |
| JGGBVFLRNNDHCM-UHFFFAOYSA-N | |
| 9673 | |
| COC(=O)C(C(F)(F)F)C(F)(F)F |
| C5H4F6O2 | |
| MFCD00054663 | |
| 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester | |
| methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate |
Specifications
| 360-54-3 | |
| 88°C | |
| MFCD00054663 | |
| 1992 | |
| JGGBVFLRNNDHCM-UHFFFAOYSA-N | |
| methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate | |
| 9673 | |
| ≥98.0% (GC) | |
| Methyl 2-(Trifluoromethyl)-3,3,3-trifluoropropionate |
| Colorless | |
| C5H4F6O2 | |
| 1 g | |
| 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester | |
| COC(=O)C(C(F)(F)F)C(F)(F)F | |
| 210.075 | |
| 210.08 | |
| Liquid |
Safety and Handling
RTECSNumber : NQ4690000
TSCA : No