CAS RN 360-54-3
IUPAC Name:
methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate
Synonyms:
methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate
Molecular Weight (g/mol):
210.08
Molecular Formula:
C5H4F6O2
InChi Key:
JGGBVFLRNNDHCM-UHFFFAOYSA-N
SMILES:
COC(=O)C(C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Methyl 2-(Trifluoromethyl)-3,3,3-trifluoropropionate 98.0+%, TCI America™
CAS: 360-54-3 Molecular Formula: C5H4F6O2 Molecular Weight (g/mol): 210.075 MDL Number: MFCD00054663 InChI Key: JGGBVFLRNNDHCM-UHFFFAOYSA-N Synonym: 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester PubChem CID: 9673 IUPAC Name: methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate SMILES: COC(=O)C(C(F)(F)F)C(F)(F)F