missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(Trifluoromethyl)-3,3,3-trifluoropropionate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 88°C |
| Quantity | 1 g |
| UN Number | 1992 |
| Formula Weight | 210.08 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-(Trifluoromethyl)-3,3,3-trifluoropropionate |
Safety and Handling
| RTECSNumber | NQ4690000 |
| TSCA | No |