missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Thiophenecarboxylate 99.0+%, TCI America™
Supplier: TCI America T11395G
Specifications
| Methyl 2-Thiophenecarboxylate | |
| Yellow | |
| C6H6O2S | |
| 5 g | |
| PGBFYLVIMDQYMS-UHFFFAOYSA-N | |
| methyl thiophene-2-carboxylate | |
| 79340 | |
| ≥99.0% (GC) |
| 5380-42-7 | |
| 98°C | |
| MFCD00016895 | |
| methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl | |
| COC(=O)C1=CC=CS1 | |
| 142.17 | |
| 142.17 | |
| Liquid |
Chemical Identifiers
| 5380-42-7 | |
| 142.17 | |
| PGBFYLVIMDQYMS-UHFFFAOYSA-N | |
| 79340 | |
| COC(=O)C1=CC=CS1 |
| C6H6O2S | |
| MFCD00016895 | |
| methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl | |
| methyl thiophene-2-carboxylate |
Safety and Handling
TSCA : No