missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Thiophenecarboxylate 99.0+%, TCI America™

Catalog No. p-7136627
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11395G 5 g
T113925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11395G Supplier TCI America Supplier No. T11395G
Only null left

Chemical Identifiers

CAS 5380-42-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD00016895
InChI Key PGBFYLVIMDQYMS-UHFFFAOYSA-N
Synonym methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl
PubChem CID 79340
IUPAC Name methyl thiophene-2-carboxylate
SMILES COC(=O)C1=CC=CS1

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 142.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Thiophenecarboxylate