missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-phenoxybenzoate, 99%

Catalog No. p-7058422
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1458409 10 g
AAL1458418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1458409 Supplier Thermo Scientific Chemicals Supplier No. L1458409

CAS: 21905-56-6 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21905-56-6
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD03093064
InChI Key PUGYLBSXMKBSRP-UHFFFAOYSA-N
PubChem CID 607606
IUPAC Name methyl 2-phenoxybenzoate
SMILES COC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Specifications

Boiling Point 130°C (0.5 mmHg)
Refractive Index 1.576
Quantity 10 g
Beilstein 2115719
Synonym 2-phenoxybenzoic acid methyl ester, benzoic acid, 2-phenoxy-, methyl ester, methyl o-phenoxybenzoate, acmc-1ce6i, #, benzoic acid,2-phenoxy-, methyl ester, benzoic acid, o-phenoxy-, methyl ester
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Methyl 2-phenoxybenzoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only