missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Oxoindoline-6-carboxylate 98.0+%, TCI America™
$70.84 - $346.71
Chemical Identifiers
| CAS | 14192-26-8 |
|---|---|
| Molecular Formula | C10H9NO3 |
| Molecular Weight (g/mol) | 191.186 |
| MDL Number | MFCD03095196 |
| InChI Key | YFTGUNWFFVDLNM-UHFFFAOYSA-N |
| Synonym | 2-Oxoindoline-6-carboxylic Acid Methyl Ester |
| PubChem CID | 3734372 |
| IUPAC Name | methyl 2-oxo-1,3-dihydroindole-6-carboxylate |
| SMILES | COC(=O)C1=CC2=C(CC(=O)N2)C=C1 |
Chemical Identifiers
| 14192-26-8 | |
| 191.186 | |
| YFTGUNWFFVDLNM-UHFFFAOYSA-N | |
| 3734372 | |
| COC(=O)C1=CC2=C(CC(=O)N2)C=C1 |
| C10H9NO3 | |
| MFCD03095196 | |
| 2-Oxoindoline-6-carboxylic Acid Methyl Ester | |
| methyl 2-oxo-1,3-dihydroindole-6-carboxylate |
Specifications
| 14192-26-8 | |
| White-Yellow | |
| MFCD03095196 | |
| 2-Oxoindoline-6-carboxylic Acid Methyl Ester | |
| COC(=O)C1=CC2=C(CC(=O)N2)C=C1 | |
| 191.186 | |
| 191.19 | |
| Crystalline Powder |
| 212°C | |
| C10H9NO3 | |
| 1 g | |
| YFTGUNWFFVDLNM-UHFFFAOYSA-N | |
| methyl 2-oxo-1,3-dihydroindole-6-carboxylate | |
| 3734372 | |
| ≥98.0% (GC) | |
| Methyl 2-Oxoindoline-6-carboxylate |
Safety and Handling
TSCA : No