accessibility menu, dialog, popup

UserName

CAS RN 14192-26-8

CH3OOOHN

CAS RN 14192-26-8

IUPAC Name: methyl 2-oxo-2,3-dihydro-1H-indole-6-carboxylate
Synonyms: methyl 2-oxo-1,3-dihydroindole-6-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: YFTGUNWFFVDLNM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2CC(=O)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results