missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxoindane-1-carboxylate, 97%

Catalog No. aah5753503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5753503 1 g
AAH5753506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5753503 Supplier Thermo Scientific Chemicals Supplier No. H5753503
Only null left

CAS: 104620-34-0 | C11H10O3 | 190.198 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.

Chemical Identifiers

CAS 104620-34-0
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD06200715
InChI Key RWSYHHRDYRDVQL-UHFFFAOYSA-N
Synonym methyl 2-oxo-1-indanecarboxylate, methyl 2-oxoindane-1-carboxylate, methyl2-oxo-1-indanecarboxylate, methyl 2-oxo-2,3-dihydro-1h-indene-1-carboxylate, 1h-indene-1-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester, acmc-20agv9, 1-methoxycarbonyl-2-indanone, 2-oxo-indan-1-carboxylic acid methyl ester, 2-oxo-1,3-dihydro-indene-1-carboxylic acid methyl ester
PubChem CID 10888732
IUPAC Name methyl 2-oxo-1,3-dihydroindene-1-carboxylate
SMILES COC(=O)C1C(=O)CC2=CC=CC=C12

Specifications

Melting Point 63°C to 70°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material Methyl 2-oxoindane-1-carboxylate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only