accessibility menu, dialog, popup

UserName

CAS RN 104620-34-0

IUPAC Name: methyl 2-oxo-2,3-dihydro-1H-indene-1-carboxylate
Synonyms: methyl 2-oxo-1,3-dihydroindene-1-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: RWSYHHRDYRDVQL-UHFFFAOYNA-N
SMILES: COC(=O)C1C(=O)CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results