missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Oxocyclopentanecarboxylate 97.0+%, TCI America™

Catalog No. C0812500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0812500ML 500 mL
C081225ML 25 mL
C0812100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0812500ML Supplier TCI America Supplier No. C0812500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10472-24-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00001411
InChI Key PZBBESSUKAHBHD-UHFFFAOYSA-N
Synonym methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane
PubChem CID 66328
IUPAC Name methyl 2-oxocyclopentane-1-carboxylate
SMILES COC(=O)C1CCCC1=O

Specifications

Color Yellow
Boiling Point 111°C
Quantity 500 mL
Formula Weight 142.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxocyclopentanecarboxylate