accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 10472-24-9

IUPAC Name: methyl 2-oxocyclopentane-1-carboxylate
Synonyms: methyl 2-oxocyclopentane-1-carboxylate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: PZBBESSUKAHBHD-UHFFFAOYNA-N
SMILES: COC(=O)C1CCCC1=O
Boiling Point 
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)