missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Oxo-1-pyrrolidineacetate 97.0+%, TCI America™

Catalog No. o02055g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02055G 5 g
O020525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02055G Supplier TCI America Supplier No. O02055G
Only null left

Chemical Identifiers

CAS 59776-88-4
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00003198
InChI Key HPDZYDTXCOFUOY-UHFFFAOYSA-N
Synonym 2-Oxo-1-pyrrolidineacetic Acid Methyl Ester, 2-Pyrrolidinone-1-acetic Acid Methyl Ester
PubChem CID 108835
IUPAC Name methyl 2-(2-oxopyrrolidin-1-yl)acetate
SMILES COC(=O)CN1CCCC1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 157.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxo-1-pyrrolidineacetate