CAS RN 59776-88-4
IUPAC Name:
methyl 2-(2-oxopyrrolidin-1-yl)acetate
Synonyms:
methyl 2-(2-oxopyrrolidin-1-yl)acetate
Molecular Weight (g/mol):
157.17
Molecular Formula:
C7H11NO3
InChi Key:
HPDZYDTXCOFUOY-UHFFFAOYSA-N
SMILES:
COC(=O)CN1CCCC1=O
1
–
1
of
1
results
1
Methyl 2-Oxo-1-pyrrolidineacetate 97.0+%, TCI America™
CAS: 59776-88-4 Molecular Formula: C7H11NO3 Molecular Weight (g/mol): 157.17 MDL Number: MFCD00003198 InChI Key: HPDZYDTXCOFUOY-UHFFFAOYSA-N Synonym: 2-Oxo-1-pyrrolidineacetic Acid Methyl Ester, 2-Pyrrolidinone-1-acetic Acid Methyl Ester PubChem CID: 108835 IUPAC Name: methyl 2-(2-oxopyrrolidin-1-yl)acetate SMILES: COC(=O)CN1CCCC1=O