missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-octynoate, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019898
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019898 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019898 Supplier Merck Emd Millipore Supplier No. 689821ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 111-12-6
Density 0.92 g/mL (at 25°C)
Flash Point 89°C - closed cup
Physical Form Neat
Linear Formula CH3(CH2)4 C///CCO2 CH3
MDL Number MFCD00009530
Boiling Point 217°C to 220°C
Synonym Methyl heptine carbonate; NSC 72098
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.21
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less