missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Nitrobenzoate 98.0+%, TCI America™

Catalog No. N015825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N015825G Supplier TCI America Supplier No. N015825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-27-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007136
InChI Key AOXPHVNMBPFOFS-UHFFFAOYSA-N
Synonym methyl o-nitrobenzoate, benzoic acid, 2-nitro-, methyl ester, 2-nitro-benzoic acid methyl ester, benzoic acid, o-nitro-, methyl ester, 2-nitrobenzoic acid methyl ester, methyl-2-nitrobenzoate, methyl o-nitro benzoate, methyl ortho-nitrobenzoate, acmc-209ml4, nitrobenzoic acid methyl ester
PubChem CID 69072
IUPAC Name methyl 2-nitrobenzoate
SMILES COC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 275°C
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Nitrobenzoate
EINECSNumber (3)-1503
TSCA Yes