missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-naphthoate, 99%

Catalog No. AAL0997303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0997303 1 g
AAL0997306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0997303 Supplier Thermo Scientific Chemicals Supplier No. L0997303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2459-25-8 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2459-25-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00017345
InChI Key IODOXLXFXNATGI-UHFFFAOYSA-N
Synonym methyl 2-naphthoate, methyl 2-naphthalenecarboxylate, 2-naphthalenecarboxylic acid, methyl ester, 2-naphthoic acid, methyl ester, 2-carbomethoxynaphthalene, 2-naphthalenecarboxylic acid methyl ester, acmc-1cliu, methyl-2-naphthoate, 2-naphthalenecarboxylicacid, methyl ester
PubChem CID 137605
IUPAC Name methyl naphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 75°C to 79°C
Boiling Point 290°C
Quantity 1 g
Beilstein 2046448
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material Methyl 2-naphthoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only