missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)benzoate, 98%

Catalog No. AAA1090506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1090506 5 g
AAA1090503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1090506 Supplier Thermo Scientific Chemicals Supplier No. A1090506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3704-28-7 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3704-28-7
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00068039
InChI Key CPQDZXPLQXZJGF-UHFFFAOYSA-N
PubChem CID 279586
IUPAC Name methyl 2-methylsulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1SC

Specifications

Melting Point 64°C to 66°C
Odor Stench
Quantity 5 g
Beilstein 2084294
Synonym methyl 2-methylthio benzoate, methyl 2-methylsulfanyl benzoate, benzoic acid, 2-methylthio-, methyl ester, benzoic acid,2-methylthio-, methyl ester, 2-methylthiobenzoic acid, methyl ester, acmc-1cnec, methyl o-methylthiosalicylate, methyl-2-methylthio benzoate, methyl 2-methylsulfanyl benzoate #, 2-methylthio benzoic acid methyl ester
Formula Weight 182.24
Percent Purity 98%
Chemical Name or Material Methyl 2- (methylthio)benzoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only