Learn More
Methyl 2-methylpropionimidate hydrochloride, 97%
CAS: 39739-60-1 | C5H12ClNO | 137.61 g/mol
Supplier: Thermo Scientific Chemicals H6338603
Description
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
| Methyl 2-methylpropionimidate hydrochloride | |
| White | |
| (CH3)2CHC(=NH)OCH3·HCl | |
| 1 g | |
| Slightly soluble in water. | |
| Cl.COC(=N)C(C)C | |
| 137.61 | |
| 137.61 | |
| Solid |
| 39739-60-1 | |
| C5H12ClNO | |
| MFCD03844704 | |
| methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1 | |
| RMXGPHAHPIIESQ-UHFFFAOYSA-N | |
| methyl 2-methylpropanimidate;hydrochloride | |
| 371709 | |
| 97% |
Chemical Identifiers
| 39739-60-1 | |
| 137.61 | |
| RMXGPHAHPIIESQ-UHFFFAOYSA-N | |
| 371709 | |
| Cl.COC(=N)C(C)C |
| C5H12ClNO | |
| MFCD03844704 | |
| methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1 | |
| methyl 2-methylpropanimidate;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only