missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-methylpropionimidate hydrochloride, 97%
Description
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Linear Formula | (CH3)2CHC(=NH)OCH3·HCl |
| Quantity | 250 mg |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 137.61 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | Methyl 2-methylpropionimidate hydrochloride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only