Learn More
Methyl 2-methylnicotinate, 97%
CAS: 65719-09-7 | C8H9NO2 | 151.165 g/mol
$161.88 - $580.78
Chemical Identifiers
| CAS | 65719-09-7 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00051961 |
| InChI Key | KLHWBYHFWALOIJ-UHFFFAOYSA-N |
| PubChem CID | 7015327 |
| IUPAC Name | methyl 2-methylpyridine-3-carboxylate |
| SMILES | CC1=C(C=CC=N1)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0907406
|
Thermo Scientific Chemicals
L0907406 |
5 g |
Each for $161.88
|
|
|||||
|
AAL0907414
|
Thermo Scientific Chemicals
L0907414 |
25 g |
Each for $580.78
|
|
|||||
Description
Methyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.
Solubility
Soluble in chloroform and methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 65719-09-7 | |
| 151.165 | |
| KLHWBYHFWALOIJ-UHFFFAOYSA-N | |
| methyl 2-methylpyridine-3-carboxylate |
| C8H9NO2 | |
| MFCD00051961 | |
| 7015327 | |
| CC1=C(C=CC=N1)C(=O)OC |
Specifications
| 65719-09-7 | |
| C8H9NO2 | |
| MFCD00051961 | |
| 471919 | |
| Soluble in chloroform and methanol. | |
| CC1=C(C=CC=N1)C(=O)OC | |
| 151.165 | |
| 151.17 | |
| Methyl 2-methylnicotinate |
| 102°C to 104°C (10 mmHg) | |
| 1.5165 | |
| 5 g | |
| methyl 2-methylnicotinate, 2-methylnicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-methyl-, methyl ester, methyl2-methylnicotinate, pubchem2218, acmc-1belg, methyi 2-methyinicotinate, methyl 2-methyinicotinate, methyl-2-methylnicotinate, ksc493g0t | |
| KLHWBYHFWALOIJ-UHFFFAOYSA-N | |
| methyl 2-methylpyridine-3-carboxylate | |
| 7015327 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only