accessibility menu, dialog, popup

UserName

CH3NOOCH3

CAS RN 65719-09-7

IUPAC Name: methyl 2-methylpyridine-3-carboxylate
Synonyms: methyl 2-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: KLHWBYHFWALOIJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)N=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results