missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylnicotinate, 97%

Catalog No. AAL0907414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0907414 25 g
AAL0907406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0907414 Supplier Thermo Scientific Chemicals Supplier No. L0907414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65719-09-7 | C8H9NO2 | 151.165 g/mol

Methyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 65719-09-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00051961
InChI Key KLHWBYHFWALOIJ-UHFFFAOYSA-N
PubChem CID 7015327
IUPAC Name methyl 2-methylpyridine-3-carboxylate
SMILES CC1=C(C=CC=N1)C(=O)OC

Specifications

Boiling Point 102°C to 104°C (10 mmHg)
Refractive Index 1.5165
Quantity 25 g
Beilstein 471919
Synonym methyl 2-methylnicotinate, 2-methylnicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-methyl-, methyl ester, methyl2-methylnicotinate, pubchem2218, acmc-1belg, methyi 2-methyinicotinate, methyl 2-methyinicotinate, methyl-2-methylnicotinate, ksc493g0t
Solubility Information Soluble in chloroform and methanol.
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material Methyl 2-methylnicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only