missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methyl-5-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133096
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22205G 5 g
M222025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22205G Supplier TCI America Supplier No. M22205G

Chemical Identifiers

CAS 77324-87-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
InChI Key TVGKPVDDCDQBRC-UHFFFAOYSA-N
Synonym methyl 5-nitro-2-methylbenzoate, 2-methyl-5-nitro-benzoic acid methyl ester, 2-methyl-5-nitrobenzoic acid methyl ester, methyl2-methyl-5-nitrobenzoate, methyl 3-nitro-6-methylbenzoate, benzoic acid, 2-methyl-5-nitro-, methyl ester, methyl 2-methyl-5-nitro-benzoate, acmc-1bix6, methyl 5-nitro-o-toluate, ksc495s6b
PubChem CID 4377758
IUPAC Name methyl 2-methyl-5-nitrobenzoate
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 69°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methyl-5-nitrobenzoate