missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methyl-4-nitrobenzoate 98.0+%, TCI America™

Catalog No. M25455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25455G 5 g
M254525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25455G Supplier TCI America Supplier No. M25455G

Chemical Identifiers

CAS 62621-09-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD03618407
InChI Key JJHCLPDHYBSSHC-UHFFFAOYSA-N
Synonym 2-Methyl-4-nitrobenzoic Acid Methyl Ester, Methyl 4-Nitro-o-toluate, 4-Nitro-o-toluic Acid Methyl Ester
PubChem CID 15088700
IUPAC Name methyl 2-methyl-4-nitrobenzoate
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 76°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methyl-4-nitrobenzoate