missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methyl-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21445G 5 g
M214425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21445G Supplier TCI America Supplier No. M21445G
Only null left

Chemical Identifiers

CAS 59382-59-1
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00082564
InChI Key CRZGFIMLHZTLGT-UHFFFAOYSA-N
Synonym benzoic acid, 2-methyl-3-nitro-, methyl ester, methyl 3-nitro-o-toluate, 2-methyl-3-nitrobenzoic acid methyl ester, methyl 3-nitro-2-methylbenzoate, 3-nitro-o-toluic acid methyl ester, 2-methyl-3-nitromethylbenzoate, pubchem10933, acmc-1awh8, ksc495s1j, methyl2-methyl-3-nitrobenzoate
PubChem CID 601165
IUPAC Name methyl 2-methyl-3-nitrobenzoate
SMILES COC(=O)C1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methyl-3-nitrobenzoate