missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-3-nitrobenzoate, 97%

Catalog No. AAH6024206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6024206 5 g
AAH6024214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6024206 Supplier Thermo Scientific Chemicals Supplier No. H6024206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59382-59-1 | C9H9NO4 | 195.17 g/mol

Methyl 2-methyl-3-nitrobenzoate may be used in the synthesis of methyl indole-4-carboxylate, 5-aminoisoquinolin-1(2H)-one, 5-nitroisocoumarin and [2-(4-fluorophenyl)-1H-indol-4-yl]-1-pyrrolidinylmethanone. It may also be used in the preparation of substituted nitrostyrene benzoic acids, via reaction with aromatic aldehydes in the presence of DBU in DMSO; and 4-(hydroxymethyl)-1-tosylindole, via Batcho-Leimgruber modification of the Reissert indole synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methyl-3-nitrobenzoate may be used in the synthesis of methyl indole-4-carboxylate, 5-aminoisoquinolin-1(2H)-one, 5-nitroisocoumarin and [2-(4-fluorophenyl)-1H-indol-4-yl]-1-pyrrolidinylmethanone. It may also be used in the preparation of substituted nitrostyrene benzoic acids, via reaction with aromatic aldehydes in the presence of DBU in DMSO; and 4-(hydroxymethyl)-1-tosylindole, via Batcho-Leimgruber modification of the Reissert indole synthesis.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 59382-59-1
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00082564
InChI Key CRZGFIMLHZTLGT-UHFFFAOYSA-N
Synonym benzoic acid, 2-methyl-3-nitro-, methyl ester, methyl 3-nitro-o-toluate, 2-methyl-3-nitrobenzoic acid methyl ester, methyl 3-nitro-2-methylbenzoate, 3-nitro-o-toluic acid methyl ester, 2-methyl-3-nitromethylbenzoate, pubchem10933, acmc-1awh8, ksc495s1j, methyl2-methyl-3-nitrobenzoate
PubChem CID 601165
IUPAC Name methyl 2-methyl-3-nitrobenzoate
SMILES COC(=O)C1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 62°C to 65°C
Quantity 5 g
Beilstein 1964020
Solubility Information Soluble in methanol.
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 2-methyl-3-nitrobenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only