missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methoxypropionate, 98%

Catalog No. AAL1352714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1352714 25 g
AAL1352706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1352714 Supplier Thermo Scientific Chemicals Supplier No. L1352714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17639-76-8 | C5H10O3 | 118.132 g/mol

Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 17639-76-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00151664
InChI Key VABBJJOSOCPYIT-UHFFFAOYSA-N
Synonym methyl 2-methoxypropionate, propanoic acid, 2-methoxy-, methyl ester, acmc-20akie, d +-alpha-methoxy-propionic acid methyl ester, methyl2-methoxypropanoate, methyl 2-methoxy-propionate, 2-methoxypropanoic acid methyl ester, racemic 2-methoxy-propionic acid methyl ester
PubChem CID 86599
IUPAC Name methyl 2-methoxypropanoate
SMILES CC(C(=O)OC)OC

Specifications

Boiling Point 43°C to 44°C (15 mmHg)
Flash Point 36°C (96°F)
Refractive Index 1.397
Quantity 25 g
UN Number UN3272
Beilstein 1720819
Solubility Information Soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl 2-methoxypropionate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 241-622-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only