accessibility menu, dialog, popup

UserName

H3COCH3OOCH3

CAS RN 17639-76-8

IUPAC Name: methyl 2-methoxypropanoate
Synonyms: methyl 2-methoxypropanoate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: VABBJJOSOCPYIT-UHFFFAOYNA-N
SMILES: COC(C)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results