missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Methoxyphenylacetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 180.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-Methoxyphenylacetate |
Safety and Handling
| TSCA | No |