missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methoxy-5-nitrobenzoate 98.0+%, TCI America™

Catalog No. M14375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14375G Supplier TCI America Supplier No. M14375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34841-11-7
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00673023
InChI Key DOBFJVVTBNTGCW-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757
PubChem CID 7020925
IUPAC Name methyl 2-methoxy-5-nitrobenzoate
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 101°C
Quantity 5 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methoxy-5-nitrobenzoate