missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methoxy-5-nitrobenzoate, 98%

Catalog No. AAB2186306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2186306 5 g
AAB2186314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2186306 Supplier Thermo Scientific Chemicals Supplier No. B2186306

CAS: 34841-11-7 | C9H9NO5 | 211.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34841-11-7
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00673023
InChI Key DOBFJVVTBNTGCW-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757
PubChem CID 7020925
IUPAC Name methyl 2-methoxy-5-nitrobenzoate
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 99°C to 102°C
Quantity 5 g
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Methyl 2-methoxy-5-nitrobenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only