missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Iodobenzoate 98.0+%, TCI America™

Catalog No. p-7131502
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
I058625G 25 g
I0586250G 250 g
2 options

Catalog No. I058625G

Supplier: TCI America I058625G

Only null left

Chemical Identifiers

CAS 610-97-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00016351
InChI Key BXXLTVBTDZXPTN-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0
PubChem CID 69132
IUPAC Name methyl 2-iodobenzoate
SMILES COC(=O)C1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 274°C
Quantity 25 g
Formula Weight 262.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Iodobenzoate