Learn More
Methyl 2-iodobenzoate, 98%
CAS: 610-97-9 | C8H7IO2 | 262.046 g/mol
Supplier: Thermo Scientific Chemicals A1320418
Description
2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.
Solubility
Insoluble in water.
Notes
Light sensitive. Store at room temperature. Incompatible with oxidizing agents.
Specifications
| Methyl 2-iodobenzoate | |
| 1.73 | |
| >110°C (230°F) | |
| 1.604 | |
| 50 g | |
| Light sensitive | |
| Insoluble in water. | |
| COC(=O)C1=CC=CC=C1I | |
| 262.046 | |
| 262.05 |
| 610-97-9 | |
| 274°C | |
| C8H7IO2 | |
| MFCD00016351 | |
| 2206859 | |
| 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0 | |
| BXXLTVBTDZXPTN-UHFFFAOYSA-N | |
| methyl 2-iodobenzoate | |
| 69132 | |
| 98% |
Chemical Identifiers
| 610-97-9 | |
| 262.046 | |
| BXXLTVBTDZXPTN-UHFFFAOYSA-N | |
| 69132 | |
| COC(=O)C1=CC=CC=C1I |
| C8H7IO2 | |
| MFCD00016351 | |
| 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0 | |
| methyl 2-iodobenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-243-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only