missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Iodo-5-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7133537
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30191G 1 g
M30195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30191G Supplier TCI America Supplier No. M30191G
Only null left

Chemical Identifiers

CAS 103440-52-4
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD12198129
InChI Key DHYMLHOXGMUIDZ-UHFFFAOYSA-N
PubChem CID 14263946
IUPAC Name methyl 2-iodo-5-methylbenzoate
SMILES COC(=O)C1=C(I)C=CC(C)=C1

Specifications

Boiling Point 105°C
Quantity 1 g
Synonym 2-iodo-5-methylbenzoic acid methyl ester, benzoic acid, 2-iodo-5-methyl-, methyl ester, benzoic acid,2-iodo-5-methyl-, methyl ester, acmc-20dzjp, methyl 6-iodo-m-toluate, methyl2-iodo-5-methylbenzoate, 4-iodo-3-methoxycarbonyltoluene, 6-iodo-m-toluic acid methyl ester, methyl 2-iodanyl-5-methyl-benzoate, methyl 2-iodo-5-methylbenzoate
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Iodo-5-methylbenzoate
TSCA No
Recommended Storage Refrigerator