missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-iodo-5-methylbenzoate, 95%

Catalog No. p-7032408
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3385303 1 g
AAH3385306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3385303 Supplier Thermo Scientific Chemicals Supplier No. H3385303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103440-52-4 | C9H9IO2 | 276.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103440-52-4
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD12198129
InChI Key DHYMLHOXGMUIDZ-UHFFFAOYSA-N
PubChem CID 14263946
IUPAC Name methyl 2-iodo-5-methylbenzoate
SMILES COC(=O)C1=C(I)C=CC(C)=C1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Synonym 2-iodo-5-methylbenzoic acid methyl ester, benzoic acid, 2-iodo-5-methyl-, methyl ester, benzoic acid,2-iodo-5-methyl-, methyl ester, acmc-20dzjp, methyl 6-iodo-m-toluate, methyl2-iodo-5-methylbenzoate, 4-iodo-3-methoxycarbonyltoluene, 6-iodo-m-toluic acid methyl ester, methyl 2-iodanyl-5-methyl-benzoate, methyl 2-iodo-5-methylbenzoate
Formula Weight 276.07
Percent Purity 95%
Chemical Name or Material Methyl 2-iodo-5-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only