Learn More
Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435900050
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 15484-46-5 , 150-19-6 | |
| 116.12 | |
| RFUCOAQWQVDBEU-UHFFFAOYSA-N | |
| 11815329 | |
| COC(=O)C(=C)CO |
| C5H8O3 | |
| MFCD12545894 | |
| methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate | |
| methyl 2-(hydroxymethyl)prop-2-enoate |
Specifications
| Methyl 2-(hydroxymethyl)acrylate | |
| 15484-46-5 , 150-19-6 | |
| 71°C to 72°C (2.0 Torr) | |
| Authentic | |
| C5H8O3 | |
| 5g | |
| methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate | |
| COC(=O)C(=C)CO | |
| 116.12 | |
| 11815329 | |
| 85% |
| Stabilized | |
| 1.1290g/mL | |
| 90°C | |
| 84% min. (GC) | |
| MFCD12545894 | |
| 1.129 | |
| RFUCOAQWQVDBEU-UHFFFAOYSA-N | |
| methyl 2-(hydroxymethyl)prop-2-enoate | |
| 0.1 wt% Monomethylether hydroquinone | |
| 116.12 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning