accessibility menu, dialog, popup

UserName

H3COOCH2OH

CAS RN 15484-46-5

IUPAC Name: methyl 2-(hydroxymethyl)prop-2-enoate
Synonyms: methyl 2-(hydroxymethyl)prop-2-enoate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: RFUCOAQWQVDBEU-UHFFFAOYSA-N
SMILES: COC(=O)C(=C)CO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)