missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-5-nitrobenzoate, 98%

Catalog No. aab2527818
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2527818 50 g
AAB2527809 10 g
AAB2527830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2527818 Supplier Thermo Scientific Chemicals Supplier No. B2527818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17302-46-4 | C8H7NO5 | 197.146 g/mol

Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 17302-46-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00024695
InChI Key UUBFELFUKFJSRD-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-5-nitrobenzoate, methyl 5-nitrosalicylate, methyl-5-nitrosalicylate, 5-nitrosalicylic acid methyl ester, benzoic acid, 2-hydroxy-5-nitro-, methyl ester, 2-hydroxy-5-nitrobenzoic acid methyl ester, methyl 5-nitro-salicylate, acmc-20a62q, methyl 5-nitro-2-hydroxybenzoate, methyl-2-hydroxy-5-nitrobenzoate
PubChem CID 87041
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

Melting Point 114°C to 117°C
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-5-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-330-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only