missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Hydroxy-3-nitrobenzoate 98.0+%, TCI America™
$32.72 - $94.33
Chemical Identifiers
| CAS | 22621-41-6 |
|---|---|
| Molecular Formula | C8H7NO5 |
| Molecular Weight (g/mol) | 197.146 |
| MDL Number | MFCD00272263 |
| InChI Key | NIBVYEHAFBEVFI-UHFFFAOYSA-N |
| Synonym | methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l |
| PubChem CID | 596904 |
| SMILES | COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O |
Chemical Identifiers
| 22621-41-6 | |
| 197.146 | |
| NIBVYEHAFBEVFI-UHFFFAOYSA-N | |
| 596904 |
| C8H7NO5 | |
| MFCD00272263 | |
| methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l | |
| COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O |
Specifications
| 22621-41-6 | |
| Yellow | |
| MFCD00272263 | |
| methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l | |
| COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O | |
| 596904 | |
| ≥98.0% (GC,T) | |
| Methyl 2-Hydroxy-3-nitrobenzoate |
| 131°C | |
| C8H7NO5 | |
| 1 g | |
| NIBVYEHAFBEVFI-UHFFFAOYSA-N | |
| 197.146 | |
| 197.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No