missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Hydroxy-3-nitrobenzoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 131°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 197.15 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 2-Hydroxy-3-nitrobenzoate |
Safety and Handling
| TSCA | No |