missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-3-nitrobenzoate, 98%

CAS: 22621-41-6 | C8H7NO5 | 197.146 g/mol

$278.92 - $985.04

Chemical Identifiers

CAS 22621-41-6
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00272263
InChI Key NIBVYEHAFBEVFI-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
PubChem CID 596904
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2502906
View Documents
Thermo Scientific Chemicals
B2502906
5 g
Each for $278.92
Only null left
 
AAB2502914
View Documents
Thermo Scientific Chemicals
B2502914
25 g
Each for $985.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22621-41-6
197.146
NIBVYEHAFBEVFI-UHFFFAOYSA-N
596904
C8H7NO5
MFCD00272263
methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O

Specifications

22621-41-6
C8H7NO5
5 g
NIBVYEHAFBEVFI-UHFFFAOYSA-N
197.146
197.15
Methyl 2-hydroxy-3-nitrobenzoate
129°C to 132°C
MFCD00272263
methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
596904
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only